siRNA Details
| Gene Symbol: RAB29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135662.1 | NP_001129134.1 | 4 | 456 | ||
| NM_001135663.1 | NP_001129135.1 | 2 | 671 | |||
| NM_001135664.1 | NP_001129136.1 | 3 | 303 | |||
| NM_003929.2 | NP_003920.1 | 4 | 557 | |||
| XM_005245569.1 | XP_005245626.1 | 4 | 455 | |||
| XM_005245570.1 | XP_005245627.1 | 4 | 508 | |||
| XM_005245571.1 | XP_005245628.1 | 4 | 433 | |||
| XM_006711605.3 | XP_006711668.1 | 3 | 199 | |||
| XM_006711606.2 | XP_006711669.1 | 3 | 163 | |||
| XM_017002748.1 | XP_016858237.1 | 3 | 458 | |||
| XM_017002749.1 | XP_016858238.1 | 3 | 436 | |||
| XM_017002750.1 | XP_016858239.1 | 3 | 368 | |||
| GenBank mRNA | AK223514.1 | - | 3 | 333 | ||
| AK303879.1 | - | 2 | 422 | |||
| AK308359.1 | - | 3 | 272 | |||
| AK313000.1 | - | 4 | 445 | |||
| BC002585.2 | - | 4 | 390 | 3140744 | ||
| BT019540.1 | - | 3 | 217 | |||
| BX647960.1 | - | 4 | 522 | |||
| CR542010.1 | - | 3 | 217 | |||
| D84488.1 | - | 3 | 257 | |||
| DA284582.1 | - | 4 | 456 | |||
| DA377295.1 | - | 3 | 258 | |||

RAB29
Genomic Map