siRNA Details
| Gene Symbol: Abcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108821.1 | NP_001102291.1 | 3 | 1573 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcd1| Gene Symbol: Abcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108821.1 | NP_001102291.1 | 3 | 1573 | ||