siRNA Details
| Gene Symbol: Irak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127555.1 | NP_001121027.1 | 5 | 874 | ||
| XM_006229598.1 | XP_006229660.1 | 5 | 857 | |||
| GenBank mRNA | BC166780.1 | - | 5 | 739 | 7934982 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irak1| Gene Symbol: Irak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127555.1 | NP_001121027.1 | 5 | 874 | ||
| XM_006229598.1 | XP_006229660.1 | 5 | 857 | |||
| GenBank mRNA | BC166780.1 | - | 5 | 739 | 7934982 | |