siRNA Details
| Gene Symbol: Taz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025748.1 | NP_001020919.1 | 2 | 440 | ||
| XM_006229599.2 | XP_006229661.1 | 3 | 831 | |||
| XM_006229600.2 | XP_006229662.1 | 4 | 622 | |||
| GenBank mRNA | BC099221.1 | - | 2 | 440 | 7387882 | |

Taz
Genomic Map