siRNA Details
| Gene Symbol: Dnase1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014223.2 | NP_001014245.2 | 4, 5 | 588 | ||
| GenBank mRNA | BC079074.1 | - | 4, 5 | 544 | 7122158 | |
| DQ116785.1 | - | 4, 5 | 588 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnase1l1| Gene Symbol: Dnase1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014223.2 | NP_001014245.2 | 4, 5 | 588 | ||
| GenBank mRNA | BC079074.1 | - | 4, 5 | 544 | 7122158 | |
| DQ116785.1 | - | 4, 5 | 588 | |||