siRNA Details
| Gene Symbol: Snx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109526.2 | NP_001102996.2 | 8 | 1008 | ||
| XM_006245736.2 | XP_006245798.1 | 7 | 1314 | |||
| XM_008767497.1 | XP_008765719.1 | 8 | 2123 | |||
| XM_008767498.1 | XP_008765720.1 | 6 | 949 | |||

Snx29
Genomic Map