siRNA Details
| Gene Symbol: Cluap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014225.1 | NP_001014247.1 | 5 | 779 | ||
| XM_006245832.2 | XP_006245894.1 | 5 | 542 | |||
| XM_008767512.1 | XP_008765734.1 | 5 | 475 | |||
| XM_008767513.1 | XP_008765735.1 | 5 | 837 | |||
| XM_008767516.1 | XP_008765738.1 | 4 | 381 | |||
| XM_008767517.1 | XP_008765739.1 | 3 | 244 | |||
| XM_008767518.1 | XP_008765740.1 | 3 | 301 | |||
| GenBank mRNA | BC079022.1 | - | 5 | 779 | 7113868 | |

Cluap1
Genomic Map