siRNA Details
| Gene Symbol: Naa60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014226.2 | NP_001014248.2 | 5, 6 | 677 | ||
| XM_008767519.1 | XP_008765741.1 | 5, 6 | 734 | |||
| XM_008767520.1 | XP_008765742.1 | 5, 6 | 713 | |||
| GenBank mRNA | AY316589.1 | - | 5, 6 | 719 | ||
| BC105304.1 | - | 5, 6 | 677 | 7191187 | ||
| FQ212054.1 | - | 5, 6 | 631 | |||

Naa60
Genomic Map