siRNA Details
| Gene Symbol: Naa60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014226.2 | NP_001014248.2 | 3 | 338 | ||
| XM_008767519.1 | XP_008765741.1 | 2 | 236 | |||
| XM_008767520.1 | XP_008765742.1 | 3 | 374 | |||
| GenBank mRNA | AY316589.1 | - | 3 | 380 | ||
| BC105304.1 | - | 3 | 338 | 7191187 | ||
| FQ212054.1 | - | 3 | 342 | |||

Naa60
Genomic Map