siRNA Details
| Gene Symbol: Fam98b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234792.2 | XP_006234854.1 | 3 | 377 | ||
| Gene Symbol: RGD1560464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_343883.6 | XP_343884.5 | 1 | 368 | ||

Fam98b
Genomic Map