siRNA Details
| Gene Symbol: BAIAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199096.1 | NP_001186025.1 | 2 | 226 | ||
| NM_001199097.1 | NP_001186026.1 | 2 | 168 | |||
| NM_001199098.1 | NP_001186027.1 | 2 | 168 | |||
| NM_001199099.1 | NP_001186028.1 | 2 | 168 | |||
| NM_001286464.1 | NP_001273393.1 | 2 | 168 | |||
| NM_003933.4 | NP_003924.2 | 2 | 276 | |||
| XM_011522728.1 | XP_011521030.1 | 2 | 771 | |||
| XM_011522729.1 | XP_011521031.1 | 2 | 771 | |||
| XM_011522730.2 | XP_011521032.1 | 2 | 771 | |||
| GenBank mRNA | AK294491.1 | - | 2 | 276 | ||
| AK294744.1 | - | 2 | 128 | |||
| AK295645.1 | - | 2 | 127 | |||
| AK299309.1 | - | 2 | 128 | |||
| AK299358.1 | - | 2 | 226 | |||
| AK315059.1 | - | 2 | 276 | |||
| AL834321.1 | - | 2 | 119 | |||
| BC104966.1 | - | 2 | 139 | 8143969 | ||
| BC112129.1 | - | 2 | 151 | 8327597 | ||
| DC318470.1 | - | 2 | 168 | |||

BAIAP3
Genomic Map