siRNA Details
| Gene Symbol: Olr1457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000526.1 | NP_001000526.1 | 1 | 298 | ||
| Gene Symbol: Olr1458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000021.1 | NP_001000021.1 | 1 | 298 | ||

Olr1457
Genomic Map