siRNA Details
| Gene Symbol: Olr1493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006609.1 | NP_001006610.1 | 750 | |||
| GenBank mRNA | AF091572.1 | - | 1 | 567 | ||
| M64387.1 | - | 750 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1493| Gene Symbol: Olr1493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006609.1 | NP_001006610.1 | 750 | |||
| GenBank mRNA | AF091572.1 | - | 1 | 567 | ||
| M64387.1 | - | 750 | ||||