siRNA Details
| Gene Symbol: CDK5R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003936.4 | NP_003927.1 | 1 | 2255 | ||
| GenBank mRNA | BC041771.1 | - | 1 | 2228 | 4818320 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDK5R2| Gene Symbol: CDK5R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003936.4 | NP_003927.1 | 1 | 2255 | ||
| GenBank mRNA | BC041771.1 | - | 1 | 2228 | 4818320 | |