siRNA Details
| Gene Symbol: Hlf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | S79820.1 | - | 2 | 139 | ||
| Gene Symbol: LOC690286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001073964.3 | XP_001073964.2 | 2 | 604 | ||
| XM_006247136.1 | XP_006247198.1 | 2 | 604 | |||

Hlf
Genomic Map