siRNA Details
| Gene Symbol: Zpbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007011.1 | NP_001007012.1 | 5 | 547 | ||
| XM_006247384.2 | XP_006247446.1 | 5 | 1677 | |||
| XM_006247385.2 | XP_006247447.1 | 5 | 1604 | |||
| GenBank mRNA | AY251605.1 | - | 5 | 539 | ||
| AY251606.1 | - | 4 | 473 | |||
| BC082007.1 | - | 5 | 547 | 7131668 | ||

Zpbp2
Genomic Map