siRNA Details
| Gene Symbol: Gabpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108841.1 | NP_001102311.1 | 1273 | |||
| XM_008768584.1 | XP_008766806.1 | 7 | 882 | |||
| XM_008768585.1 | XP_008766807.1 | 7 | 877 | |||
| XM_008768586.1 | XP_008766808.1 | 7 | 877 | |||
| XM_008768587.1 | XP_008766809.1 | 6 | 687 | |||

Gabpa
Genomic Map