siRNA Details
| Gene Symbol: Arl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108842.1 | NP_001102312.1 | 2 | 312 | ||
| XM_006248191.2 | XP_006248253.1 | 2 | 587 | |||
| XM_006248192.2 | XP_006248254.1 | 2 | 437 | |||
| XM_006248193.2 | XP_006248255.1 | 2 | 586 | |||
| GenBank mRNA | BC162029.1 | - | 2 | 433 | 9100375 | |

Arl6
Genomic Map