siRNA Details
| Gene Symbol: Olr1540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001383.1 | NP_001001383.1 | 1 | 131 | ||
| Gene Symbol: Olr1536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001353.1 | NP_001001353.1 | 1 | 131 | ||

Olr1540
Genomic Map