siRNA Details
| Gene Symbol: AP3D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261826.1 | NP_001248755.1 | 26 | 3192 | ||
| NM_003938.6 | NP_003929.4 | 24 | 3006 | |||
| XM_006722932.2 | XP_006722995.1 | 26 | 3219 | |||
| XM_017027422.1 | XP_016882911.1 | 21 | 2489 | |||
| GenBank mRNA | AB208804.1 | - | 26 | 3176 | ||
| AF002163.1 | - | 3094 | ||||
| AK094460.1 | - | 1 | 1106 | |||
| AK128692.1 | - | 1 | 1523 | |||
| AK310099.1 | - | 1 | 206 | |||
| AK315899.1 | - | 2 | 181 | |||
| U91930.1 | - | 26 | 2869 | |||

AP3D1
Genomic Map