siRNA Details
| Gene Symbol: Tigit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248323.2 | XP_006248385.1 | 3, 4 | 924 | ||
| XM_008768765.1 | XP_008766987.1 | 3, 4 | 907 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tigit| Gene Symbol: Tigit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248323.2 | XP_006248385.1 | 3, 4 | 924 | ||
| XM_008768765.1 | XP_008766987.1 | 3, 4 | 907 | |||