siRNA Details
Gene Symbol: Tigit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248323.2 | XP_006248385.1 | 4 | 1076 | ||
XM_008768765.1 | XP_008766987.1 | 4 | 1059 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tigit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248323.2 | XP_006248385.1 | 4 | 1076 | ||
XM_008768765.1 | XP_008766987.1 | 4 | 1059 |