siRNA Details
| Gene Symbol: Lmln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108843.1 | NP_001102313.1 | 4, 5 | 496 | ||
| XM_006248453.1 | XP_006248515.1 | 4, 5 | 2277 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmln| Gene Symbol: Lmln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108843.1 | NP_001102313.1 | 4, 5 | 496 | ||
| XM_006248453.1 | XP_006248515.1 | 4, 5 | 2277 | |||