siRNA Details
| Gene Symbol: Iqcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014230.1 | NP_001014252.1 | 3 | 322 | ||
| XM_006248455.2 | XP_006248517.1 | 3 | 515 | |||
| XM_006248456.2 | XP_006248518.1 | 3 | 717 | |||
| GenBank mRNA | BC087078.1 | - | 3 | 322 | 7135432 | |

Iqcg
Genomic Map