siRNA Details
| Gene Symbol: Xxylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_344027.7 | XP_344028.4 | 2, 3 | 1004 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xxylt1| Gene Symbol: Xxylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_344027.7 | XP_344028.4 | 2, 3 | 1004 | ||