siRNA Details
Gene Symbol: HIST1H2AG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC016677.1 | - | 1 | 1186 | 4342191 | |
L19778.1 | - | 1 | 1194 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HIST1H2AG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC016677.1 | - | 1 | 1186 | 4342191 | |
L19778.1 | - | 1 | 1194 |