siRNA Details
| Gene Symbol: Elavl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108848.1 | NP_001102318.1 | 2 | 160 | ||
| GenBank mRNA | BC168972.1 | - | 2 | 166 | 9034405 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elavl1| Gene Symbol: Elavl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108848.1 | NP_001102318.1 | 2 | 160 | ||
| GenBank mRNA | BC168972.1 | - | 2 | 166 | 9034405 | |