siRNA Details
| Gene Symbol: Cnpy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108852.2 | NP_001102322.1 | 1 | 94 | ||
| GenBank mRNA | BC162041.1 | - | 1 | 94 | 7100495 | |
| FQ226241.1 | - | 1 | 113 | |||
| FQ229096.1 | - | 1 | 139 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnpy4| Gene Symbol: Cnpy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108852.2 | NP_001102322.1 | 1 | 94 | ||
| GenBank mRNA | BC162041.1 | - | 1 | 94 | 7100495 | |
| FQ226241.1 | - | 1 | 113 | |||
| FQ229096.1 | - | 1 | 139 | |||