siRNA Details
| Gene Symbol: Caln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077201.1 | NP_001070669.1 | 4 | 404 | ||
| XM_006249243.2 | XP_006249305.1 | 4 | 371 | |||
| GenBank mRNA | DQ914829.1 | - | 3 | 278 | ||
| DQ914830.1 | - | 4 | 404 | |||

Caln1
Genomic Map