siRNA Details
| Gene Symbol: Piwil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108853.1 | NP_001102323.1 | 8 | 914 | ||
| XM_008769200.1 | XP_008767422.1 | 7 | 972 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Piwil1| Gene Symbol: Piwil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108853.1 | NP_001102323.1 | 8 | 914 | ||
| XM_008769200.1 | XP_008767422.1 | 7 | 972 | |||