siRNA Details
| Gene Symbol: Dpp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012205.1 | NP_001012205.1 | 3, 4 | 264 | ||
| GenBank mRNA | AY557199.1 | - | 3, 4 | 264 | ||
| DQ857324.1 | - | 3, 4 | 276 | |||
| DQ857325.1 | - | 3, 4 | 243 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpp10| Gene Symbol: Dpp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012205.1 | NP_001012205.1 | 3, 4 | 264 | ||
| GenBank mRNA | AY557199.1 | - | 3, 4 | 264 | ||
| DQ857324.1 | - | 3, 4 | 276 | |||
| DQ857325.1 | - | 3, 4 | 243 | |||