siRNA Details
| Gene Symbol: Nckap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769507.1 | XP_008767729.1 | 6 | 927 | ||
| XM_008769508.1 | XP_008767730.1 | 7 | 1191 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nckap5| Gene Symbol: Nckap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769507.1 | XP_008767729.1 | 6 | 927 | ||
| XM_008769508.1 | XP_008767730.1 | 7 | 1191 | |||