siRNA Details
| Gene Symbol: Yod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008889.1 | NP_001008889.1 | 2 | 627 | ||
| GenBank mRNA | BC107904.1 | - | 2 | 1120 | 7454897 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yod1| Gene Symbol: Yod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008889.1 | NP_001008889.1 | 2 | 627 | ||
| GenBank mRNA | BC107904.1 | - | 2 | 1120 | 7454897 | |