siRNA Details
| Gene Symbol: Ikbke | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108854.1 | NP_001102324.1 | 15 | 1988 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ikbke| Gene Symbol: Ikbke | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108854.1 | NP_001102324.1 | 15 | 1988 | ||