siRNA Details
Gene Symbol: Ncf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100984.1 | NP_001094454.1 | 2 | 237 | ||
XM_006250013.2 | XP_006250075.1 | 2 | 450 | |||
XM_008769664.1 | XP_008767886.1 | 2 | 447 | |||
GenBank mRNA | JN864041.1 | - | 2 | 237 | ||
JN864042.1 | - | 2 | 237 | |||
JN864045.1 | - | 2 | 237 | |||
JN864046.1 | - | 2 | 237 |
