siRNA Details
| Gene Symbol: Ncf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100984.1 | NP_001094454.1 | 12 | 1138 | ||
| XM_006250013.2 | XP_006250075.1 | 12 | 1330 | |||
| XM_008769664.1 | XP_008767886.1 | 10 | 1102 | |||
| GenBank mRNA | JN864041.1 | - | 11 | 1135 | ||
| JN864042.1 | - | 11 | 1135 | |||
| JN864045.1 | - | 11 | 1129 | |||
| JN864046.1 | - | 10 | 1021 | |||

Ncf2
Genomic Map