siRNA Details
Gene Symbol: Ncf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250013.2 | XP_006250075.1 | 10, 11 | 1202 | ||
XM_008769664.1 | XP_008767886.1 | 8, 9 | 974 | |||
GenBank mRNA | JN864041.1 | - | 9, 10 | 1007 | ||
JN864042.1 | - | 9, 10 | 1007 | |||
JN864045.1 | - | 9, 10 | 1001 | |||
JN864046.1 | - | 8, 9 | 893 |
