siRNA Details
| Gene Symbol: Rgsl2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198790.1 | NP_942085.1 | 1 | 303 | ||
| GenBank mRNA | AJ515383.1 | - | 1 | 303 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgsl2h| Gene Symbol: Rgsl2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198790.1 | NP_942085.1 | 1 | 303 | ||
| GenBank mRNA | AJ515383.1 | - | 1 | 303 | ||