siRNA Details
| Gene Symbol: RGD1561635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769692.1 | XP_008767914.1 | 1, 2 | 245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561635| Gene Symbol: RGD1561635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769692.1 | XP_008767914.1 | 1, 2 | 245 | ||