siRNA Details
| Gene Symbol: Slamf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191550.1 | NP_001178479.1 | 4 | 706 | ||
| XM_006250249.2 | XP_006250311.1 | 4 | 920 | |||
| XM_006250250.2 | XP_006250312.1 | 4 | 918 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slamf7| Gene Symbol: Slamf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191550.1 | NP_001178479.1 | 4 | 706 | ||
| XM_006250249.2 | XP_006250311.1 | 4 | 920 | |||
| XM_006250250.2 | XP_006250312.1 | 4 | 918 | |||