siRNA Details
| Gene Symbol: RERE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042681.1 | NP_001036146.1 | 16 | 2426 | ||
| NM_001042682.1 | NP_001036147.1 | 6 | 725 | |||
| NM_012102.3 | NP_036234.3 | 17 | 2611 | |||
| XM_005263464.2 | XP_005263521.1 | 16 | 2365 | |||
| XM_005263466.1 | XP_005263523.1 | 10 | 1248 | |||
| XM_011541510.1 | XP_011539812.1 | 15 | 2351 | |||
| XM_011541511.1 | XP_011539813.1 | 16 | 2477 | |||
| XM_017001358.1 | XP_016856847.1 | 16 | 2116 | |||
| XM_017001359.1 | XP_016856848.1 | 16 | 2621 | |||
| GenBank mRNA | AB007927.2 | - | 15 | 1761 | ||
| AB036737.1 | - | 17 | 2437 | |||
| AF016005.1 | - | 6 | 703 | |||
| AF118275.1 | - | 2 | 139 | |||

RERE
Genomic Map