siRNA Details
| Gene Symbol: LOC305806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013993.1 | NP_001014015.1 | 486 | |||
| GenBank mRNA | BC079250.1 | - | 486 | 7133060 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC305806| Gene Symbol: LOC305806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013993.1 | NP_001014015.1 | 486 | |||
| GenBank mRNA | BC079250.1 | - | 486 | 7133060 | ||