siRNA Details
| Gene Symbol: RERE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042681.1 | NP_001036146.1 | 22, 23 | 5286 | ||
| NM_001042682.1 | NP_001036147.1 | 12, 13 | 3585 | |||
| NM_012102.3 | NP_036234.3 | 23, 24 | 5471 | |||
| XM_005263464.2 | XP_005263521.1 | 22, 23 | 5225 | |||
| XM_005263466.1 | XP_005263523.1 | 16, 17 | 4108 | |||
| XM_011541510.1 | XP_011539812.1 | 21, 22 | 5211 | |||
| XM_017001358.1 | XP_016856847.1 | 22, 23 | 4976 | |||
| XM_017001359.1 | XP_016856848.1 | 22, 23 | 5481 | |||
| GenBank mRNA | AB007927.2 | - | 21, 22 | 4621 | ||
| AB036737.1 | - | 23, 24 | 5297 | |||
| AF016005.1 | - | 12, 13 | 3563 | |||
| AF118275.1 | - | 8, 9 | 2999 | |||

RERE
Genomic Map