siRNA Details
| Gene Symbol: Tmem55b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014233.2 | NP_001014255.1 | 3 | 486 | ||
| XM_006251888.1 | XP_006251950.1 | 3 | 459 | |||
| XM_006251889.2 | XP_006251951.1 | 3 | 451 | |||
| GenBank mRNA | BC087604.1 | - | 3 | 489 | 7312219 | |
| FQ225412.1 | - | 3 | 464 | |||

Tmem55b
Genomic Map