siRNA Details
| Gene Symbol: Rnase12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012209.1 | NP_001012209.1 | 1 | 50 | ||
| XM_006251892.2 | XP_006251954.1 | 2 | 230 | |||
| XM_008770557.1 | XP_008768779.1 | 2 | 1736 | |||
| XM_008770558.1 | XP_008768780.1 | 2 | 242 | |||

Rnase12
Genomic Map