siRNA Details
| Gene Symbol: Rnase1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029904.1 | NP_001025075.1 | 737 | |||
| GenBank mRNA | J00771.1 | - | 2 | 731 | ||
| Gene Symbol: LOC103690354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770630.1 | XP_008768852.1 | 2 | 746 | ||

Rnase1
Genomic Map