siRNA Details
| Gene Symbol: Rnf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108868.1 | NP_001102338.2 | 6, 7 | 912 | ||
| XM_006252000.2 | XP_006252062.1 | 6, 7 | 1613 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf31| Gene Symbol: Rnf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108868.1 | NP_001102338.2 | 6, 7 | 912 | ||
| XM_006252000.2 | XP_006252062.1 | 6, 7 | 1613 | |||