siRNA Details
| Gene Symbol: Rnf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108868.1 | NP_001102338.2 | 20 | 3187 | ||
| XM_006252000.2 | XP_006252062.1 | 20 | 3885 | |||
| GenBank mRNA | BC169074.1 | - | 12 | 1425 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf31| Gene Symbol: Rnf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108868.1 | NP_001102338.2 | 20 | 3187 | ||
| XM_006252000.2 | XP_006252062.1 | 20 | 3885 | |||
| GenBank mRNA | BC169074.1 | - | 12 | 1425 | ||