siRNA Details
| Gene Symbol: Cideb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108869.1 | NP_001102339.1 | 3, 4 | 468 | ||
| XM_006252002.2 | XP_006252064.1 | 3, 4 | 511 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cideb| Gene Symbol: Cideb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108869.1 | NP_001102339.1 | 3, 4 | 468 | ||
| XM_006252002.2 | XP_006252064.1 | 3, 4 | 511 | |||