siRNA Details
| Gene Symbol: Arl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013433.1 | NP_001013451.1 | 2 | 347 | ||
| XM_006252135.2 | XP_006252197.1 | 3 | 1114 | |||
| XM_008770766.1 | XP_008768988.1 | 3 | 1109 | |||
| GenBank mRNA | BC091183.1 | - | 2 | 347 | 7365828 | |

Arl11
Genomic Map